Electronic Torque Wrenches
Ergonomically designed molded non-slip rubber grip
The COMPUTORQ3 Electronic Torque Wrench is a simple to use digital readout wrench that displays real time torque values in any of four torque units, ft. lbs., in. lbs., Nm, kg. cm.
Simply set the desired torque value and apply force until the green LED illuminates. Great for light industrial, automotive, motorcycle, watercraft, and aircraft applications.
* Sleek new design
* 15 degree flex ratchet in head
* Push button socket release head
* Large, easy to read digital display
* Long battery life, uses two each CR123 lithium batteries
* Torques in both clockwise and counterclockwise directions
* Battery saving auto-shutoff after two minutes
* COMPUTORQ3 remembers last unit and torque setting selected
* 3 color LED light bar
* When desired torque value is reached, display will hold value and
$919.00 - $1,100.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-ft)
Availability Pricing
J. H. Williams Tool Group 1002CF3 Torque Wrench, 3/8" drive, 10 to 100" pounds EW-10780-43 Mfr # 1002EFRMH
COMPARE
100
$1,100.00USD / Each
J. H. Williams Tool Group 2401CI3 Torque Wrench, 1/4" drive, 2.0 to 20.0 foot pounds EW-10780-47 Mfr # 2401EFRMH
COMPARE
20
$919.00USD / Each