Cole-Parmer® CN-200 Series Digital Heating Controller
Get accurate temperature control of heating equipment in your lab
  • Three-digit LED display allows you to set a 1°C resolution over a range of –10 to 800°C
  • Front-panel up/down controls make for easy programming
  • Ideal for heating mantles or or heating tapes
Versatile benchtop digital heating controller can be used in three ways: in on/off mode with the hysteresis loop controlling power switching, as a PID controller, or simply as a temperature measuring device using a probe. Sample temperatures are displayed on an easy-to-read LED display. Use the included PTFE-covered thermometer probe for measurements up to 250°C or order optional temperature probe 32587-98 for measuring up to 800°C. There is a 5-pin DIN socket on the controller for the probe.
Rugged zinc die-cast outer housing is suitable for benchtop use or wall mounting. Rear clamp accepts a standard 1/2" (12.5 mm) rod to mount on a support stand or frame.
$1,140.00 - $1,168.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Control Output 1 Description
Availability Pricing
COMPARE
230 1500 W Digital Heating Controller, 1500 W; 230 VAC
$1,140.00USD / Each
COMPARE
115 750 W Digital Heating Controller, 750 W; 115 VAC
$1,168.00USD / Each