Electrothermal Electric Bunsen Burners
Provides clean and easy operation
  • Top cowl deflects heat away from hands
  • Air circulation from the vented housing keeps base cool to the touch
  • Efficient — only consumes 400 W of power
This electric Bunsen burner combines the advantages of a regular gas burner with the clean and easy operation of a heating mantle. Corrosion-resistant stainless steel burner features a cone-shaped coiled resistance heater which directs radiant heat out from the cavity. Cooling vents circulate air to cool the base, making it possible to hold while in use. A detachable cowl also deflects heat from the medium surface.
Ideal for noncontact radiant heating of materials in test tubes, crucibles, small flasks, and beakers. Effective heating of a test tube sample should be done 25 mm above the top of the burner. Crucible heating may be achieved by removing the cowl.
Note: To regulate the power input and temperature of the Bunsen burner, a heating controller is required. Without a controller, the burner operates continuously at full heating power. The Bunsen burner can be purchased with or without the recommended controller (order controller separately).
$702.50 - $1,237.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
230 Electric Bunsen Burner, without Controller; 230 V
$702.50USD / Each
COMPARE
230 Electric Bunsen Burner, with Controller; 230 V
$1,237.50USD / Each
COMPARE
115 Electric Bunsen Burner, with Controller; 115 V
$1,237.50USD / Each
COMPARE
115 Electric Bunsen Burner, without Controller; 115 V
$1,020.00USD / Each