This product is now discontinued, View Related Products
Cole-Parmer® MP-250D Digital Melting Point Apparatus
Height adjustable extension arm ensures proper viewing height for each user
  • View up to three samples simultaneously for greater efficiency
  • Integral light and 8x magnifying lens enhance sample observation
  • Digital processor provides fast warm up and accurate temperature control
  • Store up to four melt temperatures in temporary memory
  • Extension arm folds away for easy storage
This ergonomically designed digital melting point apparatus offers convenience and comfort at an affordable price. An adjustable extension arm can be set to different viewing heights making it ideal for multiple users. A rotating viewing head and safety eye piece eliminate glare on the lens and protect you against hot zone. Conveniently located push button controls allow temperature recording without looking away from the sample.
To use, simply load your samples, key in your set point temperature and once the oven temperature reaches the set point, audible beeps indicate oven temperature stability. The temperature then slowly ramps up from the set point and the melt is observed through the 8x magnifying lens; select 1°C/min or 10°C/min ramp rate. View melt temperature on the LCD. An ECO mode saves on energy use when keypad remains unused for a specified length of time.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
1° or 10°C per minute (selectable) 400 240