Environmental Express BOD Bottle Accessories
Using a fresh, clean bottle for each analysis provides more accurate results in your lab
  • Ideal for 300 mL BOD bottles
The use of a cap over the mouth of a the BOD bottle to prevent evaporation is recommended by Standard Methods. Our overcaps are made of polyethylene and fit snugly against the rim and shoulder of our disposable BOD bottles. These overcaps do not fit the glass BOD bottles.
The acrylic stoppers work with all brands of glass and plastic 300 mL BOD bottles.
$49.20USD / Pkg of 50
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Environmental Express Overcaps for BOD Bottles, PE; 50/Pk
$49.20USD / Pkg of 50