Environmental Express® MicroBlock Snip & Pour Pre-Measured Reagents
Deliver precise volume with pre-measured reagents
  • Easy-to-use, just snip and pour into receiver tube
  • Increase accuracy by eliminating the need to measure and pipette
Pre-measured Snip & Pour reagent tubes for Ammonia and Cyanide Distillations deliver the precise volume of reagent to each sample — working perfectly with the MicroBlock™ systems!
Snip & Pour Reagents are packaged in convenient, chemical-resistant PTFE tubes. Reagent addition is a snap with Snip & Pour reagent tubes. Just snip the corner of the PTFE tube and pour the contents directly into the receiver tube.
Each pre-measured PTFE Tube for MicroBlock delivers 10mL of reagent.
$109.50 - $115.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (mL) Description
Availability Pricing
COMPARE
7664-93-9 500 0.2N H2SO4, 50 x 10 mL/Pk
$109.50USD / Each
COMPARE
1310-73-2 500 1.25N NaOH, 50 x 10 mL/Pk
$115.00USD / Each
COMPARE
10043-35-3 500 20g/L Boric Acid, 50 x 10 mL/Pk
$109.50USD / Each