Environmental Express® SimpleDist Snip & Pour Reagents
Reagent addition is a snap with Snip & Pour reagent tubes
  • Eliminate measuring and pipetting
  • Increase accuracy
  • Improve quality control and final results
  • Meet DOT requirements
Snip & Pour concentrates are packaged in convenient PTFE tubes. Just snip the corner of the PTFE tube and pour the contents directly into SimpleDist® collection trap. Premeasured reagents are meant for use with cyanide or ammonia distillation.
$88.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
SimpleDist® Snip & Pour Reagent, 0.2 N H2SO4, 5 mL Tube; 50/PK
$88.50USD / Each
COMPARE
SimpleDist® Snip & Pour Reagent, 1.25 N NaOH, 5 mL Tube; 50/PK
$88.50USD / Each
COMPARE
SimpleDist® Snip & Pour Reagent, 20 g/L Boric Acid, 5 mL Tube; 50/PK
$88.50USD / Each