Environmental Express® Snip & Pour® Standards
Premeasured controls for oil and grease analysis
  • Meets requirements for EPA Method 1664
  • Each lot is quality checked gravitmetrically and by GC
$111.32 - $136.58USD / Pkg of 20
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
40 mg HEM in 10 mL Acetone; 20/Pk
$111.32USD / Pkg of 20
COMPARE
40 mg HEM in 5 mL Acetone; 20/Pk
$122.01USD / Pkg of 20
COMPARE
120 mg HEM in 30 mL Acetone; 9/Pk
$136.58USD / Pkg of 20
COMPARE
7 mg HEM in 10 mL Acetone; 20/Pk
$122.04USD / Pkg of 20
COMPARE
5 mg HEM in 10 mL Acetone; 20/Pk
$118.46USD / Pkg of 20
COMPARE
10 mg HEM in 10 mL Acetone; 20/Pk
$122.01USD / Pkg of 20