Environmental Express StepSaver Oil and Grease Extraction
Solid phase extraction systems simplify EPA Method 1664 for oil and grease
  • Ready to be evaporated without a separate drying step
StepSaver systems streamline the extraction process by allowing the analyst to collect the solvent with eluted oil and grease directly into a collection flask. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. It is ready to be evaporated without a separate drying step.
$1,364.75 - $7,204.85USD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
System, 1-Station, PVC/Glass; Ea
$1,539.85USD / Each
COMPARE
System, 3-Station, PVC/Glass; Ea
$3,599.85USD / Each
COMPARE
System, 7-Station, PVC/Glass; Ea
$6,998.85USD / Each
COMPARE
System, 1-Station, SS/Glass; Ea
$1,848.85USD / Each
COMPARE
System, 3-Station, SS/Glass; Ea
$4,011.85USD / Each
COMPARE
System, 6-Station, SS/Glass; Ea
$7,101.85USD / Each
COMPARE
System, 7-Station, PVC/UltraFlow; Ea
$5,690.75USD / Each
COMPARE
System, 1-Station, SS/UltraFlow; Ea
$1,673.75USD / Each
COMPARE
System, 6-Station, SS/UltraFlow; Ea
$5,999.75USD / Each
COMPARE
System, 1-Station, PVC/UltraPrep; Ea
$1,467.75USD / Each
COMPARE
System, 3-Station, PVC/UltraPrep; Ea
$3,115.75USD / Each
COMPARE
System, 7-Station, PVC/UltraPrep; Ea
$5,793.75USD / Each
COMPARE
System, 3-Station, SS/UltraPrep; Ea
$3,424.75USD / Each
COMPARE
System, 6-Station, SS/UltraPrep; Ea
$6,102.75USD / Each
COMPARE
System, 1-Station, PVC UltraPrep; Glass Funnel
$1,642.85USD / Each
COMPARE
System, 3-Station, PVC UltraPrep, Glass Funnel
$3,702.85USD / Each
COMPARE
System, 7-Station, PVC UltraPrep, Glass Funnel
$7,101.85USD / Each
COMPARE
System, 1-Station, SS, UltraPrep, Glass Funnel
$1,951.85USD / Each
COMPARE
System, 3-Station, SS, UltraPrep, Glass Funnel
$4,114.85USD / Each
COMPARE
System, 6-Station, SS, UltraPrep, Glass Funnel
$7,204.85USD / Each
Showing 1 - 20 of 24