Environmental Express® StirBase Magnetic Stirrer
Turn your existing HotBlock into a stirring HotBlock to meet EPA Method 3060A
  • A 48 MGO Neodymium Iron Boron drive magnet powers this extra-large surface stirrer
  • Stirring positions are only limited by the number of vessels you can place on the plate surface
  • Remote controller allows for safe operation when used in a fume hood
  • Custom-designed stir bars mix samples vertically to ensure complete mixing
The StirBase magnetic stirrer can be used with most HotBlock and HotBlock Pro systems, or as a stand-alone stirrer with unlimited stirring positions. The detached controller allows for 100 different stirring speed settings with a maximum of 2150 rpm and has a 6-ft (1.8-m) cable for safe operation of the stirrer in a fume hood. The stirrer housing is corrosion-resistant PVC.
An extremely strong internal magnet allows you to place an entire HotBlock on the platform and provide stirring to each well.* Sixty 7/8" long x 1/2" diameter (23.1 x 11.5 mm) stir bars are included and fit in the 50- and 100-mL digestion cups, as well as many other laboratory vessels. Stir bars are PEEK encapsulated for superior chemical resistance and compatibility.
The StirBase stirrer cannot be used around aluminum or magnetic items as it will cause drag on the motor, and or the maximum stir speed cannot be achieved and the motor will overheat. Additionally, stir bars should be placed in the mixing vessel prior to being placed on the stirrer.
$6,839.00 - $14,999.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
$14,999.00USD / Each
COMPARE
240
$6,839.00USD / Each