Eppendorf 5810R Series Multi-Purpose Refrigerated Centrifuges
Versatile rotor and adapter options can be used for virtually any application!
  • Superior quality and metal safety containment construction with aerosol tight lid options ensures your safety
  • Software control guarantees high accuracy, speed and temperature for optimal performance
Use these high-speed, multi-purpose centrifuges for a variety of applications: cell harvesting, high-speed separation of cell lysates, and DNA precipitations. The automatic rotor sensing feature will recognize your rotors and detect imbalances for safe operation. Refrigerator units cool rotor chamber in about 15 minutes and keep chambers at a constant temperature while lid is down, even at maximum speeds. Set the temperature range between 9°C and 40°C. The built-in drain eliminates water accumulated from condensation and prevents corrosion. Order rotors and adapters separately.
Model 5810R accept up to 4 x 500 mL bottles, 28 x 50 mL conical tubes, or 16 x microplates — ideal for high throughput labs needing flexibility in sample running.
$11,756.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Eppendorf 022625501 Multi-purpose centrifuge 5810R; 4 x 500 mL capacity, 120 VAC EW-02570-03 Mfr # 022625501
COMPARE
115
$11,756.00USD / Each
Eppendorf 022625551 Multi-purpose centrifuge 5810R; 4 x 500 mL capacity, 230 VAC EW-02570-04 Mfr # 022625551
COMPARE
230
$11,756.00USD / Each