Erlab Captair Ductless Filtering Fume Hoods and Weighing Stations
Simpler to use, safer to operate
  • A true analysis of your chemical handlings by our in-house laboratory chemists
  • A certificate of validation for chemicals used in the hood
  • Complies with the AFNOR NFX 15-211 standard (Class 1 and 2)
  • Chemical Questionnaire completion required
The ductless filtering fume hood and powder weighing stations are designed with advanced carbon filtration to provide you with a safe working environment. Chemical exposure is eliminated by the configurable filtering system above the enclosure. Installation is simple and no ductwork is necessary allowing for relocation. The units will arrive completely knocked down and will require assembly.
Configurable filter columns allow for complete versatility, providing the ability to be configured to handle gases, solvents, acids, powders, and particulates. Audible and visual signals alert you of the product performance, such as face velocity, static pressure, and filter efficiency. One push of a button to activates the system. The hoods come with Smart Technology™ which includes intuitive light-based communication interface to keep you constantly informed of the level of safety. An added value is the on-board eGuard web-based program, monitoring of the system, which can be done remotely using your PC or mobile app.
Options such as a hinged sloped sash design, or reverse sash are available. Order work surface and base separately. Filters are ordered separately, based on your handlings and validation report and needed for product functionality.
$5,195.00 - $13,350.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Description
Availability Pricing
Erlab Captair 321 Ductless Filtering Fume Hood, 30.75", Angled Sash; 80 to 220 VAC EW-33520-01 Mfr # 321SMART
COMPARE
30.75 Ductless Fume Hood, 32\" W, angled sash; 80 to 220 VAC
$5,195.00USD / Each
Erlab Captair 391 Ductless Filtering Fume Hood, 39.5", Angled Sash; 80 to 220 VAC EW-33520-03 Mfr # 391SMART
COMPARE
39.375 Ductless Fume Hood, 39\" W, angled sash; 80 to 220 VAC
$6,175.00USD / Each
Erlab Captair 481 Ductless Filtering Fume Hood, 49.6", Angled Sash; 80 to 220 VAC EW-33520-05 Mfr # 481SMART
COMPARE
49.625 Ductless Fume Hood, 49\" W, angled sash; 80 to 220 VAC
$7,850.00USD / Each
Erlab Captair 483 Ductless Filtering Fume Hood, 50, Vertical Sash; 80 to 220 VAC EW-33520-06 Mfr # 483SMART
COMPARE
50 Ductless Fume Hood, 50\" W, vertical sash; 80 to 220 VAC
$9,650.00USD / Each
Erlab Captair 633 Ductless Filtering Fume Hood, 63", Vertical Sash; 80 to 220 VAC EW-33520-07 Mfr # 633SMART
COMPARE
63 Ductless Fume Hood, 63\" W, vertical sash; 80 to 220 VAC
$11,400.00USD / Each
Erlab Captair 714 Ductless Filtering Fume Hood, 71", Vertical Sash; 80 to 220 VAC EW-33520-11 Mfr # 714SMART
COMPARE
70.875 Ductless Fume Hood, 71\" W, vertical sash; 80 to 220 VAC
$13,350.00USD / Each