Erlab AirStore Ductless Filtering Chemical Storage Cabinets
Simpler to use, safer to operate
  • Audible and visual alerts warn you when there is a problem or malfunction
  • Mobile app lets you monitor and control conditions remotely
  • Single-button activation begins air purification quickly and easily
The ductless filtering chemical storage cabinets are designed with advanced carbon filtration and clear lockable doors for easy secure viewing of contents. The cabinets are filtered 24 hours, 7 days a week. You will not find a build up of vapors or odors present within the cabinet as the air is continuously being turned over. No HVAC connection to the outside required. Simply press a button to activate the system or remotely control and monitor your systems from anywhere using a PC or the free eGuard app. Each storage cabinet comes with Smart Technology™ which includes intuitive light-based communication interface to keep you constantly informed of the level of safety.
Available storage configurations include large-volume pull-out drawers, sliding doors, hinged doors with shelves, and pull-out doors with trays. FIlters sold separately.
$2,850.00 - $5,260.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (in) Power (VAC) Width (in)
Availability Pricing
Erlab AirStore 822 B Ductless Filtering Chemical Storage Cabinets with Sliding Doors, 29" W; 80 to 220 VAC EW-33520-13 Mfr # 822SMARTV1
COMPARE
29 3/4 85 to 240 25 9/16
$2,850.00USD / Each
Erlab AirStore 834 Ductless Filtering Chemical Storage Cabinets with Double Doors, 35" W; 80 to 220 VAC EW-33520-15 Mfr # 834SMART1V1
COMPARE
90 9/16 85 to 240 35 7/16
$5,260.00USD / Each