This product is now discontinued, View Related Products
ESS Precleaned EPA Narrow-Mouth HDPE Bottles
High-density polyethylene (HDPE) bottles are precleaned according to EPA procedure 3 methods for metal, cyanide, sulfide, and fluoride analysis.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Outside Diameter (OD) (in)
Availability Pricing
ESS PC 0250-1000 Bottle, Narrow-Mouth, HDPE, 8 oz, 24/cs EW-99535-60 Mfr # PC 0250-1000
COMPARE
250 High Density Polyethyene (HDPE) 2 1/8
Call For Price
ESS PC 0500-1000 Bottle, Narrow-Mouth, HDPE, 16 oz, 12/cs EW-99535-61 Mfr # PC 0500-1000
COMPARE
500 High Density Polyethyene (HDPE) 2 5/8
Call For Price
ESS PC 0950-1000 Bottle, Narrow-Mouth, HDPE, 32 oz, 12/cs EW-99535-62 Mfr # PC 0950-1000
COMPARE
1000 High Density Polyethyene (HDPE) 3 3/8