This product is now discontinued, View Related Products

Ethylenediamine Anhydrous

  • Assay - 98-101%
  • Water - ≤2.0%
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical E4794 Ethylenediamine Anhydrous (Certified) (4 liter) EW-88073-23 Mfr # E4794
COMPARE
107-15-3
Call For Price
Fisher Chemical E479500 Ethylenediamine Anhydrous (Certified) (500mL) EW-88073-24 Mfr # E479500
COMPARE
107-15-3
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat