This product is now discontinued, View Related Products

Fisher Chemical Ethylenediamine Tetraacetic Acid

Quality and consistency your lab can count on
  • Chelating agent for metal ions
  • Minimizes metal ion impurities in reaction buffers
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Fisher Chemical BP118500 Ethylenediaminetetraacetic Acid (White Cryst. Powd.) (500g) EW-88056-72 Mfr # BP118500
COMPARE
60-00-4 500 Ethylenediaminetetraacetic Acid (White Cryst. Powd.) (500g)
Call For Price
Fisher Chemical BP24821 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (1 liter) EW-88059-54 Mfr # BP24821
COMPARE
60-00-4 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (1 liter)
Call For Price
Fisher Chemical BP2482100 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (100mL) EW-88059-55 Mfr # BP2482100
COMPARE
60-00-4 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (100mL)
Call For Price
Fisher Chemical BP248220 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (20 liter) EW-88059-56 Mfr # BP248220
COMPARE
60-00-4 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (20 liter)
Call For Price
Fisher Chemical BP2482500 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (500mL) EW-88059-57 Mfr # BP2482500
COMPARE
60-00-4 Ethylenediaminetetraacetic Acid (0.5 M Solution/pH 8.0) (500mL)
Call For Price
Fisher Chemical E47810 Ethylenediamine Tetraacetic Acid (Certified ACS) (10kg) EW-88073-22 Mfr # E47810
COMPARE
60-00-4 Ethylenediamine Tetraacetic Acid (Certified ACS) (10kg)
Call For Price
COMPARE
60-00-4 500 EDTA, Free Acid, ACS Grade, 500 g
Call For Price
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat