This product is now discontinued, View Related Products
Extech EasyView Light Meter
Measures up to 20,000 lux in four ranges
Color- and cosine-corrected light meter measures up to 20,000 lux in four ranges and up to 20,000 footcandles in four ranges. Readings are simultaneously displayed numerically and in a bar graph. Meter is available with or without NIST-traceable calibration report supplied by the manufacturer.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Certifications
Availability Pricing
Extech EasyView Light Meter EW-01588-05 Mfr # EA31
COMPARE
Call For Price
Extech EA31-NIST Easyview Light Meter with NIST Calibration Certificate EW-01588-07 Mfr # EA31-NIST
COMPARE
NIST-Traceable
Call For Price
Extech EA31 Easyview 31 Digital Light Meter EW-50510-91 Mfr # EA31
COMPARE
None