This product is now discontinued, View Related Products
Extech EC400 Conductivity/TDS/Salinity Pocket Testers
Waterproof meter for use in waste, boiler; cooling water; pure and drinking water
  • Autoranging with 8 selectable units: µS, mS, ppm, ppt, mg/L, g/L, °C, and °F
  • Analog bargraph indicates trends
  • Data Hold, Auto Power Off, Low battery indicator
  • Simultaneous display of conductivity or TDS plus Temperature
  • Internal memory stores up to 15 labeled readings for easy recall
  • Adjustable Conductivity to TDS ratio factor from 0.4 to 1.0
  • Self calibration on power up
  • Waterproof to IP67
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Conductivity Max Salinity Max TDS
Availability Pricing
Extech EC400 conductivity/TDS/salinity tester EW-53026-12 Mfr # EC400
COMPARE
Call For Price
Extech EC410 conductivity/TDS/salinity tester kit EW-53026-14 Mfr # EC410