Extech EX360 Series CAT IV True RMS Multimeters with Noncontact Voltage Detection
Maximize worker safety with CAT IV 600 V rating and noncontact voltage detector
  • Protection against 8000 V peak impulse transient voltage allows safe usage at utility connection points
  • Noncontact voltage detection with LED indicator confirms a circuit is live or not before testing
  • LoZ feature prevents false readings from ghost voltages
  • Smart Data Hold freezes reading, and alerts audibly and visually if measurement changes ±50 counts
  • Choose from models with features specific to three applications — electrical, HVAC, or industrial
True RMS provides accurate AC measurements. Additional features include peak hold to capture the highest reading, relative mode for capacitance zero and offset adjustment, and auto power-off (with disable).
ITEM(S) ON SALE
$100.29USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Cable Identifier AC Max (Volts) Description
Availability Pricing
Extech EX360 Electrical MultiMeter with Non-Contact Voltage Detector EW-20026-07 Mfr # EX360
COMPARE
±0.5% 1000 Electrical MultiMeter with Non-Contact Voltage Detector
$100.29USD / Each
$117.99USD / Each