Extech Heavy-Duty Industrial Multimeters
Double-molded IP67-rated design provides waterproof protection
  • Rugged design withstands drops up to 6 feet
  • Meters are rated CAT IV 600 V
These heavy-duty industrial multimeters provide 1000 V input protection on all functions and a diode open circuit voltage of 2.8 VDC. Additional features include dual sensitivity frequency functions, auto power-off with disable feature, min/max, relative, data hold, and a large backlit LCD display with bar graph for easy operator interpretation.
Model 26852-20 features resistance, continuity, diode, frequency and duty cycle measurements, autoranging or manual range selection, and a 6000-count LCD display.
Model 26852-22 offers all of the functions of model 26852-20, and includes True-RMS measuring capabilities plus capacitance, and a beaded-wire type K probe.
Model 26852-24 offers all the functions of both models plus peak hold, 4 to 20 mA% measurement, a 44,000-count LCD display for high resolution, and precision accuracy of ±0.06%.
Model 26852-48 offers all the functions of models 26852-20, -22, and includes data acquisition mode for real-time data transmission to your PC.
Model 26852-49 offers all the functions of models 26852-20, -22, 20034-48, and includes a built-in noncontact infrared thermometer with laser pointer.
ITEM(S) ON SALE
$163.19 - $233.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) Resistance Max (MΩ) True Rms
Availability Pricing
Extech EX520 True RMS Industrial Multimeter EW-26852-22 Mfr # EX520
COMPARE
1000 60 True RMS
$163.19USD / Each
$191.99USD / Each
Extech EX530 Precision True RMS Industrial Multimeter EW-26852-24 Mfr # EX530
COMPARE
1000 40 True RMS
$233.99USD / Each