Extech MN30 True RMS AutoSensing™ Multimeter with NCV
Automatically recognizes proper measurement mode
  • Obtain True RMS readings up to 600 V AC/DC
  • Quickly detect voltage with built-in noncontact voltage (NCV) detector
  • Safely see in dark places—features LED flashlight
  • Compact size easily fits in your pocket or toolbox
The unique AutoSensing™ feature of the MN30 digital multimeter automatically changes modes for voltage or resistance readings. Additional features include a flashlight, audible continuity tester, overload and low-battery indication, and auto-off power—all with a CAT III-600 V safety rating.
$43.00 - $55.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) Ac Voltage Accuracy DC Max (Volts)
Availability Pricing
Extech MN30 TRMS AutoSensing Multimeter + NCV EW-20000-79 Mfr # MN30
COMPARE
600 ±(1.8% 3) 600
$43.00USD / Each
Extech MN36 MULTIMETERMINIAUTORANGING EW-26046-36 Mfr # MN36
COMPARE
$55.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat