Extech WTH600 Wireless Weather Station Kit
Don't get caught in the rain
  • Provides indoor comfort level and outdoor weather readings
  • High/Low alarms for temperature, wind speed, and rainfall
  • Transmits at 915 MHz; FCC approved for use in North America
This complete personal weather station allows you to plan activities such as yard work and gardening, exercise and recreation, or field work and outdoor research. Forecast shows current conditions—sunny, partly cloudy, cloudy, rainy, stormy, or snowy. Bar chart shows historical data for rainfall or barometric pressure. Base unit measures indoor temperature and humidity and indicates comfort level via icon. Amber LED indicators flash during alarm or backlight the display to indicate low battery or signal strength. Radio-controlled atomic clock automatically synchronizes for accuracy, but that feature can be disabled if necessary. Transmission range up to 450 ft (150 m) line of sight between transmitter and base.
Model 35500-07 (WTH600) transmits at 915 MHz and is FCC approved for use in North America. Model 35725-11 (WTH600-E) transmits at 868 MHz for use in Europe; all other regions require manual clock setup.
ITEM(S) ON SALE
$149.59USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description Max Temperature (Inside) (° F) Max Temperature (Outside) (° C)
Availability Pricing
Extech WTH600-KIT Wireless Weather Station EW-35500-07 Mfr # WTH600-KIT
COMPARE
Wireless Weather Station 158 80
$149.59USD / Each
$175.99USD / Each