Extra-Strength Pleated Cellulose Cartridges
Pleated filter gives you more surface area for higher flowrate and longer life.
THis cellulose cartridge is blended with polyester for better wet strength. The polyester material allows more pleats to improve flow and dirt loading. Suitable to 165°F (74°C).
Note: For optimal performance, change cartridge at 25 psid.
$12.90 - $13.40USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
COMPARE
10 15 GPM 5
$13.40USD / Each
COMPARE
10 15 GPM 1
$12.90USD / Each