This product is now discontinued, View Related Products

Fiberboard Mailer Narrow-Mouth HDPE bottle

Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Qorpak 3300 Narrow-Mouth Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs EW-06148-23 Mfr # PLC-03390
COMPARE
Narrow-Mouth Bottle Mailer, 2 oz, 4-1/2\" L x 1-3/4\" dia, 24/cs
Call For Price
Qorpak 3301 Narrow-Mouth Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs EW-06148-33 Mfr # PLC-03403
COMPARE
Narrow-Mouth Bottle Mailer, 4 oz, 5-1/2\" L x 2\" dia, 24/cs
Call For Price
Qorpak 3302 Narrow-Mouth Bottle Mailer, 8 oz, 6-1/2" L x 2-1/2" dia, 24/cs EW-06148-43 Mfr # PLC-03412
COMPARE
Narrow-Mouth Bottle Mailer, 8 oz, 6-1/2\" L x 2-1/2\" dia, 24/cs
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat