This product is now discontinued, View Related Products

Fiberboard Mailer with medium round glass bottle

Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Qorpak 2304 Medium Round Glass Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs EW-06148-15 Mfr # GLC-01465
COMPARE
Medium Round Glass Bottle Mailer, 1 oz, 3-3/4\" L x 1-1/2\" dia, 24/cs
Call For Price
Qorpak 2305 Medium Roung Glass Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs EW-06148-25 Mfr # GLC-01492
COMPARE
Medium Roung Glass Bottle Mailer, 2 oz, 4-1/2\" L x 1-3/4\" dia, 24/cs
Call For Price
Qorpak 2306 Medium Round Glass Bottle Mailer, 4 oz, 5-1/2" L x 2" dia, 24/cs EW-06148-35 Mfr # GLC-01522
COMPARE
Medium Round Glass Bottle Mailer, 4 oz, 5-1/2\" L x 2\" dia, 24/cs
Call For Price
Qorpak 2307 Medium Round Glass Bottle Mailer, 8 oz, 6-1/2" L x 2-1/2" dia, 24/cs EW-06148-45 Mfr # GLC-01551 Y
COMPARE
Medium Round Glass Bottle Mailer, 8 oz, 6-1/2\" L x 2-1/2\" dia, 24/cs
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat