Fiberboard Mailer with wide-mouth HDPE bottle

Meet government regulations for mailing liquids
Safely ship nonhazardous samples with these mailing tubes — each mailing tube is designed to securely hold one bottle. Fiberboard tubes feature metal bottoms and screw-on caps. Order labels separately.
$259.00 - $264.00USD / Case of 24

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Qorpak 3303 Wide Mouth Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs
Buy More Save More
EW-06148-14 Mfr # PLC-03558
COMPARE
Wide Mouth Bottle Mailer, 1 oz, 3-3/4" L x 1-1/2" dia, 24/cs
$259.00USD / Case of 24
Qorpak 3304 Wide-Mouth Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs
Buy More Save More
EW-06148-24 Mfr # PLC-03565
COMPARE
Wide-Mouth Bottle Mailer, 2 oz, 4-1/2" L x 1-3/4" dia, 24/cs
$264.00USD / Case of 24
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat