Fisher BioReagents™ Triton X-100® Wetting Agent, Electrophoresis Grade
Quality and consistency your lab can count on
  • This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$94.90 - $128.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Packaging Volume (mL) Description
Availability Pricing
Fisher BioReagents Triton X-100® Wetting Agent, Electrophoresis Grade, 100 mL EW-88057-48 Mfr # BP151100
COMPARE
Glass Bottle 100 Triton X-100® Wetting Agent, Electrophoresis Grade, 100 mL
$94.90USD / Each
Fisher BioReagents Triton X-100® Wetting Agent, Electrophoresis Grade, 500 mL EW-88057-49 Mfr # BP151500
COMPARE
HDPE Bottle 500 Triton X-100® Wetting Agent, Electrophoresis Grade, 500 mL
$128.50USD / Each