Fisher Chemical Chloroform
Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$310.00 - $547.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Fisher Chemical C6064 Chloroform (Approx. 0.75% Ethanol as Preservative/HPLC) (4 liter) EW-88005-28 Mfr # C6064
COMPARE
HPLC 4000 Chloroform (Approx. 0.75% Ethanol as Preservative/HPLC) (4 L)
$310.00USD / Each
Fisher Chemical C6074 Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 liter) EW-88005-34 Mfr # C6074
COMPARE
HPLC 4000 Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 L)
$360.00USD / Each
Fisher Chemical C607SK4 Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 liter) EW-88069-91 Mfr # C607SK4
COMPARE
HPLC 4000 Chloroform (Approx. 50ppm Pentene as Preservative/HPLC) (4 L)
$547.00USD / Each