Fisher Chemical™ o-Phosphoric Acid, 85%

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$283.00 - $442.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Fisher Chemical o-Phosphoric Acid, 85%, Certified ACS Grade, 2.5 L EW-88019-44 Mfr # A242212
COMPARE
Certified ACS 2500 o-Phosphoric Acid, 85%, Certified ACS Grade, 2.5 L
$442.50USD / Each
Fisher Chemical o-Phosphoric Acid, 85%, HPLC Grade, 500 mL EW-88051-11 Mfr # A260500
COMPARE
HPLC 500 o-Phosphoric Acid, 85%, HPLC Grade, 500 mL
$283.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat