This product is now discontinued, View Related Products

Fisher Chemical™ Potassium Bromide, Infrared Grade

Consistent, high quality organics and fine chemicals at great savings
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Fisher Chemical Potassium Bromide, Infrared Grade, 25 g EW-88019-76 Mfr # P22725
COMPARE
7758-02-3 25 Potassium Bromide, Infrared Grade, 25 g
Call For Price
Fisher Chemical P205500 Potassium Bromide (Beads or Powder/Certified ACS) (500g) EW-88019-78 Mfr # P205500
COMPARE
7758-02-3 500 Potassium Bromide (Beads or Powder/Certified ACS) (500g)
Call For Price
Fisher Chemical P2053 Potassium Bromide (Beads or Powder/Certified ACS) (3kg) EW-88019-80 Mfr # P2053
COMPARE
7758-02-3 Potassium Bromide (Beads or Powder/Certified ACS) (3kg)
Call For Price
Acros Organics AC196480010 Potassium bromide 98% (1kg) CAS 7758-02-3 EW-88209-63 Mfr # AC196480010
COMPARE
7758-02-3 1000 Potassium bromide 98% (1kg) CAS 7758-02-3
Call For Price
Acros Organics AC206391000 Potassium bromide, IR grade 99+% (100g) CAS 7758-02-3 EW-88220-93 Mfr # AC206391000
COMPARE
7758-02-3 100 Potassium bromide, IR grade 99+% (100g) CAS 7758-02-3
Call For Price
Acros Organics Potassium bromide, reagent ACS (500g) CAS 7758-02-3 EW-88362-32 Mfr # AC424075000
COMPARE
7758-02-3 500 Potassium bromide, reagent ACS (500g) CAS 7758-02-3
Call For Price
COMPARE
7758-02-3 500 Potassium Bromide Crystal Acs 500g
Call For Price
COMPARE
7758-02-3 2500 Potassium Bromide Crystal Acs 2.5kg
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat