Fisher Chemical Sodium Dichromate Dihydrate

$155.00 - $526.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Fisher Chemical S235500 Sodium Dichromate Dihydrate (Crystalline/Certified ACS) (500g) EW-88023-92 Mfr # S235500
COMPARE
7789-12-0 500 Sodium Dichromate Dihydrate (Crystalline/Certified ACS) (500g)
$526.00USD / Each
Fisher Chemical S234500 Sodium Dichromate Dihydrate (Granular/Technical) (500g) EW-88080-33 Mfr # S234500
COMPARE
7789-12-0 500 Sodium Dichromate Dihydrate (Granular/Technical) (500g)
$397.00USD / Each
Acros Organics AC219240010 Sodium dichromate dihydrate 99% (1kg) CAS 7789-12-0 EW-88234-50 Mfr # AC219240010
COMPARE
7789-12-0 1000 Sodium dichromate dihydrate 99% (1kg) CAS 7789-12-0
$155.00USD / Each
COMPARE
7789-12-0 500 Sodium Dichromate 2-Hyd Tech, 500 g
$208.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat