Fisher Chemical™ Sodium Hydroxide

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$78.90 - $760.50USD / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Weight (g) Description
Availability Pricing
Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 500 g EW-88024-06 Mfr # S318500
COMPARE
Certified ACS 500 Sodium Hydroxide, Certified ACS Grade, 500 g
$158.25USD / Each
Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 1 kg EW-88024-08 Mfr # S3181
COMPARE
Certified ACS 1000 Sodium Hydroxide, Certified ACS Grade, 1 kg
$276.25USD / Each
Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 3 kg EW-88024-10 Mfr # S3183
COMPARE
Certified ACS 3000 Sodium Hydroxide, Certified ACS Grade, 3 kg
$421.00USD / Each
Fisher Chemical Sodium Hydroxide, BP/EP/FCC/JP/NF Grade, 2.5 kg EW-88024-12 Mfr # S392212
COMPARE
BP/EP/FCC/JP/NF 2500 Sodium Hydroxide, BP/EP/FCC/JP/NF Grade, 2.5 kg
$565.00USD / Each
Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 10 kg EW-88080-81 Mfr # S31810
COMPARE
Certified ACS 10000 Sodium Hydroxide, Certified ACS Grade, 10 kg
$760.50USD / Each
Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 100 g EW-88080-82 Mfr # S318100
COMPARE
Certified ACS 100 Sodium Hydroxide, Certified ACS Grade, 100 g
$78.90USD / Each
Fisher Chemical Sodium Hydroxide, Certified ACS Grade, 5 kg EW-88080-83 Mfr # S3185
COMPARE
Certified ACS 5000 Sodium Hydroxide, Certified ACS Grade, 5 kg
$581.00USD / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat