Fisher Chemical Tween 20

Quality and consistency your lab can count on
  • Hydroxyl Number - 96 to 108
  • Water - ≤3.0%
  • Residue after Ignition - ≤0.25%
  • Arsenic - ≤0.0003%
  • Lead - ≤0.0005%
  • Copper - ≤0.0005%
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$57.50 - $67.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher Chemical BP337100 Tween 20 (100 mL) CAS 9005-64-5 (Poly Bottle) EW-88065-31 Mfr # BP337100
COMPARE
9005-64-5 100 Tween 20 (100 mL) CAS 9005-64-5 (Poly Bottle)
$57.50USD / Each
Fisher Chemical BP337500 Tween 20 (500 mL) CAS 9005-64-5 (Poly Bottle) EW-88065-43 Mfr # BP337500
COMPARE
9005-64-5 500 Tween 20 (500 mL) CAS 9005-64-5 (Poly Bottle)
$67.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat