Fixed and Portable Transit-Time Flowmeters
Unique features make these flowmeters extremely advanced!
  • Signal strength analyzer
  • Anti-noise deflector technology
  • Fluid control technology
  • Cross-correlation signal detection
  • Includes Reynolds number calculation
  • Feature large, backlit LCD and simple installation
QStar™ transit-time flowmeters feature a quick setup for easy use and a large, 320 x 240 pixel LCD. All units have integrated heat quantity measurement capabilities that enables rapid and convenient recording of heat flows. Software is preprogrammed for temperature measurement. Models 32600-00, -01, and -02 also feature temperature measurement for energy readings.
•Anti-Noise Deflection Technology — Qstar’s transducers are designed to minimize the noise of ringing from the transducer and pipe, which causes disruptive signals that could interfere with the desired signal and a loss in energy for the “ringing” instead of the main signal.
•Automatic Fluid Control Technology — If the sonic velocity of the fluid changes, such as changes in temperature or in the composition of the fluid, the position of the transducer is no longer correct and the reading of the flow will change and have an additional error. QStar meters measure the sonic velocity and by refraction of the signal path continuously calculates the “miss-positioning” of the transducers and compensates for the effect of the miss-position.
•Cross Correlation Signal Detection — Signal noise can come from various sources, such as frequency converters for pumps, pipe noise, or signal scattering from particles or gas bubbles. QStar meters implement advanced signal coding and detection algorithms to digitally filter unwanted “noise” and only the correct coded signal will be identified as a valid signal from the flowmeter.
$714.00 - $8,934.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Output Description
Availability Pricing
Great Plains Industries QME05 Energy-Measuring Fixed Ultrasonic Flowmeter, 0.5 MHz EW-32620-00 Mfr # QME05
COMPARE
Max Flow Rate = 98 ft per second Energy-Measuring Fixed Ultrasonic Flowmeter, 0.5 MHz
$8,700.00USD / Each
Great Plains Industries QME10 Energy-Measuring Fixed Ultrasonic Flowmeter, 1 MHz EW-32620-01 Mfr # QME10
COMPARE
Max Flow Rate = 98 ft per second Energy-Measuring Fixed Ultrasonic Flowmeter, 1 MHz
$6,424.00USD / Each
Great Plains Industries QME20 Energy-Measuring Fixed Ultrasonic Flowmeter, 2 MHz EW-32620-02 Mfr # QME20
COMPARE
Max Flow Rate = 98 ft per second Energy-Measuring Fixed Ultrasonic Flowmeter, 2 MHz
$6,491.00USD / Each
Great Plains Industries QMF05 Fixed Ultrasonic Flowmeter, 98 ft/second, PVC, 0.5 MHz EW-32620-03 Mfr # QMF05
COMPARE
Max Flow Rate = 98 ft per second Fixed Ultrasonic Flowmeter, 0.5 MHz
$7,983.00USD / Each
Great Plains Industries QMF10 Fixed Ultrasonic Flowmeter, 1 MHz; 1/Each EW-32620-04 Mfr # QMF10
COMPARE
Max Flow Rate = 98 ft per second Fixed Ultrasonic Flowmeter, 1 MHz
$5,708.00USD / Each
Great Plains Industries QMF20 Fixed ultrasonic flowmeter, 2 MHz (2x 4-20 mA, 1 pulse, 1 relay, 1 uUSB) EW-32620-05 Mfr # QMF20
COMPARE
Max Flow Rate = 98 ft per second Fixed ultrasonic flowmeter, 2 MHz (2x 4-20 mA, 1 pulse, 1 relay, 1 uUSB)
$5,775.00USD / Each
Great Plains Industries QMP10 Portable Ultrasonic Flowmeter, 98 ft/second, Aluminum, 1 MHz EW-32620-07 Mfr # QMP10
COMPARE
Max Flow Rate = 98 ft per second Portable Ultrasonic Flowmeter, 1 MHz
$8,934.00USD / Each
Great Plains Industries QMP20 Portable Ultrasonic Flowmeter, 98 ft/second, Aluminum, 2 MHz EW-32620-08 Mfr # QMP20
COMPARE
Max Flow Rate = 98 ft per second Portable Ultrasonic Flowmeter, 2 MHz
$8,934.00USD / Each
Great Plains Industries QMF-PT1 Temperature sensor kit, for fixed flowmeters; 16 ft (4.8 m) L cable EW-32620-80 Mfr # QMF-PT1
COMPARE
For fixed flowmeters Temperature sensor kit, for fixed flowmeters; 16 ft (4.8 m) L cable
$714.00USD / Each
Great Plains Industries QMP-PT1 Temperature sensor kit, for portable flowmeters; 16 ft (4.8 m) L cable EW-32620-81 Mfr # QMP-PT1
COMPARE
For portable flowmeters Temperature sensor kit, for portable flowmeters; 16 ft (4.8 m) L cable
$714.00USD / Each
Showing 1- 10 of 10