Flexible Impeller Self-Priming Metallic Pumps
Pumps fluids in applications where durability is needed—the 12 VDC models are ideal for remote/portable use
  • Pharmaceutical—processing lab solutions, lotions, preparations, and filtering solutions
  • Industrial—circulating and transferring corrosive liquids, transferring solutions with entrained air (such as soaps and detergents), sampling, and pilot plant installations
  • Agricultural—transferring liquid fertilizers, herbicides, and pesticides
  • Metal plating—filtration of plating solutions, filling, and emptying vats
These flexible impeller self-priming metallic pumps are available in moderate-flow and moderate-flow utility options.
Moderate-flow pumps feature wetted parts of bronze and neoprene/nitrile are ideal for saltwater applications. Model 07036-10 has an 8-ft (2.4-m) three-wire cord with plug and a 3/4" GHT(M) adapter. Model 75150-10 features a permanent-magnet motor with 10" (25.4 cm) leads. Model 75150-00 has a thermal overload protected, universal motor with 6-ft (1.8 m) grounded plug.
Moderate-flow utility pumps are ideal for light commercial and industrial applications where intermittent flow is needed. Designed for 15 minutes of use and 45 minutes being off. Rugged handle for easy portability. Models include18" (45.7 cm) pigtail leads.
$173.30 - $879.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Wetted Materials Description
Availability Pricing
COMPARE
5.8 Bronze, Neoprene, 316 Stainless Steel Moderate-Flow, flexible impeller pump, neoprene, 5.8 GPM, 17.3 psi, 115 VAC
$879.50USD / Each
COMPARE
6.7 Bronze, Nitrile, 416 Stainless Steel Moderate-Flow, flexible impeller pump, bronze/nitrile, 6.7 GPM, 18 psi, 115 VAC
$187.60USD / Each
COMPARE
6.4 Bronze, Nitrile, 416 Stainless Steel Moderate-Flow, flexible impeller pump, bronze/nitrile, 6.4 GPM, 18 psi, 12 VDC
$194.00USD / Each
COMPARE
5 Plated-Brass, Nitrile Moderate-Flow Pony Utility Pump for Intermittent Use, Nickel Plated Brass, 12VDC
$173.30USD / Each
COMPARE
5 304 Stainless Steel, Nitrile Moderate-Flow Pony Utility Pump for Intermittent Use, Stainless Steel, 12VDC
$220.30USD / Each