FLIR® MIO-Series I/O Modules for Thermal Cameras
Monitor up to seven thermal imaging cameras remotely
  • Easily configure each channel for alarm or analog outputs
  • 4 to 20 mA output provides greater process control options
  • Built-in web server simplifies setup
Turn your FLIR AX8, A310, and FC-Series R thermal imaging cameras into a complete remote condition/process monitoring and fire detection system with the compact MIO series intelligent I/O modules. Easy to set up, the I/O modules are simple to configure via a built-in web server—no need for a separate PC or custom computer software.
Up to seven cameras can be connected to the I/O modules. Systems can be expanded by adding more I/O modules to the network. Once connected, the module can target designated regions of interest and start monitoring. The module can be programmed through the web server for real-time audible and visual alarms when temperature thresholds are breached. The MIO series can also output an analog 4 to 20 mA current loop signal per each region of interest, providing you with various process control options.
$1,009.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Number of Channels Description
Availability Pricing
Flir MIO-AX8-1 I/O module for AX8 Thermal Cameras EW-39757-24 Mfr # MIO-AX8-1
COMPARE
2 I/O module for AX8 Thermal Cameras
$1,009.00USD / Each