FLIR® TRMS Digital Multimeters
Diagnostic functions provide fast and effective troubleshooting
  • Automatically select the optimal measuring range with autoranging feature
  • Bright, large 6000-count 3.8” digital backlit LCD
  • Durable IP40 design for rugged applications
  • CAT III-600 V, CAT IV-300 V safety rating
These True RMS digital multimeters feature diagnostic measurements including 10 A AC/DC current, 600 V AC/DC voltage, and capacitance. Plus, you’ll work more efficiently with advanced features such as diode testing, continuity check, non-contact voltage detection, and min/max/average hold functions.
These multimeters are CAT III 600 V and CAT IV 300 V rated and can withstand a drop test up to 2 meters. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically on the large, backlit LCD with 5 reading per second update rate. Min/Max and average readings are recorded to capture variations.
$163.00 - $188.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) AC Max (Volts) Description
Availability Pricing
Flir DM62 TRMS Digital Multimeter with Non-Contact Voltage EW-35725-21 Mfr # DM62
COMPARE
$163.00USD / Each
Flir DM64 HVAC TRMS Digital Multimeter with Non-Contact Voltage and Temperature EW-35725-22 Mfr # DM64
COMPARE
$188.00USD / Each
Flir DM66 TRMS Digital Multimeter with Non-Contact Voltage and Temperature EW-35725-23 Mfr # DM66
COMPARE
$188.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat