FLIR® VS290 Thermal MSX® Videoscope
Designed to help professionals quickly and safely find hidden dangers in difficult locations
  • Troubleshoot problems from a distance with the 160 × 120 true thermal imager and 2 MP visual camera
  • Easily maneuver the 6.6-ft (2-m) long camera probe
  • Recognize the location of a temperature issue with FLIR MSX, which extracts details from the built-in visual camera and superimposes them onto the full thermal image
  • IP67-rated camera tip and IP54-rated base unit/probe provide a high level of protection against dust and water
  • Conduct electrical inspections safely with a CAT IV 600 V rating
  • Color alarms (isotherms) quickly identify areas of concern
Featuring a 160 × 120 true thermal imager and FLIR MSX® (Multi-Spectral Dynamic Imaging), the VS290 thermal videoscope enables you to catch hot spots before a failure occurs to maintain uptime. The 6.6-ft (2-m) long camera probe makes it possible to easily inspect through small openings, improving productivity and reducing diagnostic time. The probe is ideal for inspection of underground electrical distribution vaults, inside large gearboxes, motors, attics, crawl spaces, and other industrial applications. The camera probe can be easily replaced in the field so no time is lost. Data storage and transfer capabilities communicate potential issues before they become major problems. Save images and videos using the included SD memory card, and upload to a PC via USB-C cable.
$2,189.00 - $5,459.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flir Thermal MSX® Videoscope Kit with Rectangular Probe Tip EW-23029-04 Mfr # VS290-32
COMPARE
Thermal MSX® Videoscope Kit with Rectangular Probe Tip
$5,459.00USD / Each
Flir Videoscope Display and Battery for VS290 Series (No Probes) EW-23029-05 Mfr # VS290-00
COMPARE
Videoscope Display and Battery for VS290 Series (No Probes)
$2,189.00USD / Each