Aalborg® Flow Controllers for Universal Flow Control Systems
All models include a calibration certificate to NIST-traceable standards. Flow rates are unaffected by modest temperature and pressure variations—providing excellent response and accuracy Nonclogging straight-tube sensor ensures laminar gas flow for more accurate measurements
  • All models include a calibration certificate to NIST-traceable standards.
  • Flow rates are unaffected by modest temperature and pressure variationsproviding excellent response and accuracy
  • Nonclogging straight-tube sensor ensures laminar gas flow for more accurate measurements
This flow controller may be paired with an existing control system, but has been designed to be used with the Mass Flow Command Modules in Series 32681-XX.
This controller element includes both a flowmeter element and electromagnetically actuated control valve. The flowmeter design features an advanced straight-tube thermal dispersion sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet.
The meter allows for a four-point calibration across the flow range to improve meter linearity. An NIST-traceable calibration certificate is included for each controller unit.
Each controller is factory calibrated for use with a specific gas. One key benefit of this design is its strong repeatability characteristics, making it ideal for use in applications where the variability of multiple controllers can affect the quality of a process.
Controller is easily connected to the command module using an 8-ft long flat cable with integrated multipin D-sub connector (included). To protect from wiring errors, each controller is protected from shorting or polarity reversal by a resettable fuse.
Wetted parts for the controller are 316 SS and Viton® O-rings. Buna-N, Kalrez® or Neoprene O-rings are available upon request.
These controllers operate and maintain the stated full-scale accuracy and linearity at inlet pressures between 10 and 60 psi (0.68 to 4.10 bars) and gas temperatures between 59 and 77°>F (15 to 25°>C). If operating outside 10 to 60 psi, add ±0.01%/psi full-scale (±0.145fi/bar); if operating outside 59 to 77°>F, add ±0.08%/°>F full-scale (±0.15%/°>C).
$1,418.00 - $1,835.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Gas Type Max Flow Rate (SCCM) Description
Availability Pricing
Aalborg GFCS-011293 Mass flow controller, universal COMM, for Air/N2/CO, 0 to 10 sccm
Buy More Save More
EW-32668-00 Mfr # GFCS-011293
COMPARE
Air, Nitrogen, Carbon Monoxide 10 Mass Flow Controller, Universal COMM, for Air/N2/CO, 0 to 10 sccm
$1,418.00USD / Each
Aalborg GFCS-011545 Mass flow controller, universal COMM, for Air/N2/CO, 0 to 100 sccm
Buy More Save More
EW-32668-06 Mfr # AFCS-010004
COMPARE
Air, Nitrogen, Carbon Monoxide 100 Mass Flow Controller, Universal COMM, for Air/N2/CO, 0 to 100 sccm
$1,418.00USD / Each
Aalborg AFCS-010001 Mass flow controller, universal COMM, for Air/N2/CO, 0 to 200 sccm
Buy More Save More
EW-32668-08 Mfr # AFCS-010001
COMPARE
Air, Nitrogen, Carbon Monoxide 200 Mass Flow Controller, Universal COMM, for Air/N2/CO, 0 to 200 sccm
$1,418.00USD / Each
Aalborg AFCS-010007 Mass flow controller, universal COMM, for Air/N2/CO, 0 to 1 sL/min
Buy More Save More
EW-32668-12 Mfr # AFCS-010007
COMPARE
Air, Nitrogen, Carbon Monoxide 1000 Mass Flow Controller, Universal COMM, for Air/N2/CO, 0 to 1 sL/min
$1,418.00USD / Each
Aalborg AFCS-010010 Mass flow controller, universal COMM, for Air/N2/CO, 0 to 15 sL/min
Buy More Save More
EW-32668-20 Mfr # AFCS-010010
COMPARE
Air, Nitrogen, Carbon Monoxide 15000 Mass Flow Controller, Universal COMM, for Air/N2/CO, 0 to 15 sL/min
$1,835.00USD / Each
Aalborg AFCS-010022 Mass flow controller, universal COMM, for O2, 0 to 1 sL/min
Buy More Save More
EW-32669-12 Mfr # AFCS-010022
COMPARE
Oxygen 1000 Mass Flow Controller, Universal COMM, for O2, 0 to 1 sL/min
$1,418.00USD / Each
Aalborg AFCS-010055 Mass flow controller, universal COMM, for H2, 0 to 1 sL/min
Buy More Save More
EW-32676-12 Mfr # AFCS-010055
COMPARE
Hydrogen 1000 Mass Flow Controller, Universal COMM, for H2, 0 to 1 sL/min
$1,418.00USD / Each
Aalborg AFCS-010068 Mass flow controller, universal COMM, for He, 0 to 500 sccm
Buy More Save More
EW-32677-10 Mfr # AFCS-010068
COMPARE
Helium 500 Mass Flow Controller, Universal COMM, for He, 0 to 500 sccm
$1,418.00USD / Each
Aalborg AFCS-010083 Mass flow controller, universal COMM, for CO2, 0 to 500 sccm
Buy More Save More
EW-32678-10 Mfr # AFCS-010083
COMPARE
Carbon Dioxide 500 Mass Flow Controller, Universal COMM, for CO2, 0 to 500 sccm
$1,418.00USD / Each
Aalborg AFCS-010190 Mass flow controller, universal COMM, for CO2, 0 to 1 sL/min
Buy More Save More
EW-32678-12 Mfr # AFCS-010190
COMPARE
Carbon Dioxide 1000 Mass Flow Controller, Universal COMM, for CO2, 0 to 1 sL/min
$1,418.00USD / Each
Showing 1- 10 of 10