Flow Indicators and Sensors
Indicators feature clear windows to visualize and indicate positive flow. Indicators do not feature an output. Choose an indicator with high-visibility orange rotor for darker fluids.
Sensors/Switches feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25 Hz at low flow to 225 Hz at high flow.
$66.00 - $385.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Pipe Size Description
Availability Pricing
COMPARE
5 1/4" NPT(F) Flow Indicator; Polypropylene, 1/4" NPT(F), Orange Rotor
$66.00USD / Each
COMPARE
20 1/2" NPT(F) Flow Indicator; Polypropylene, 1/2" NPT(F), Orange Rotor
$66.00USD / Each
COMPARE
5 1/4" NPT(F) Flow Indicator; Brass, 1/4" NPT(F), Orange Rotor
$172.00USD / Each
COMPARE
20 1/2" NPT(F) Flow Indicator; Brass, 1/2" NPT(F), Orange Rotor
$172.00USD / Each
COMPARE
30 3/4" NPT(F) Flow Indicator; Brass, 3/4" NPT(F), Orange Rotor
$172.00USD / Each
COMPARE
30 1" NPT(F) Flow Indicator; Brass, 1" NPT(F)
$172.00USD / Each
COMPARE
5 1/4" NPT(F) Flow Indicator; Polypropylene, 1/4" NPT(F)
$130.00USD / Each
COMPARE
20 1/2" NPT(F) Flow Indicator; Polypropylene, 1/2" NPT(F)
$130.00USD / Each
COMPARE
5 1/4" NPT(F) Flow Indicator; Brass, 1/4" NPT(F)
$243.00USD / Each
COMPARE
20 1/2" NPT(F) Flow Indicator; Brass, 1/2" NPT(F)
$243.00USD / Each
COMPARE
20 1/2" NPT(F) Flow Sensor/Switch; Polypropylene, 1/2" NPT(F), 24 VDC
$243.00USD / Each
COMPARE
5 1/4" NPT(F) Flow Sensor/Switch; Brass, 1/4" NPT(F), 24 VDC
$326.00USD / Each
COMPARE
30 1" NPT(F) Flow Sensor/Switch; Brass, 1" NPT(F), 24 VDC
$385.00USD / Each
Showing 1- 13 of 13