Flowline Buoyancy Vertical-Mount Level Switch
Don’t let your tanks overflow
  • Optional strobe alarm alerts you to high level conditions
  • Fail-safe relay control of pumps and valves with 0 to 60 second time delay
  • Buoyancy switches are ideal for clean water and noncoating chemicals
  • NEMA 4X rated enclosure with 300° conduit swivel
  • Select length for custom set points
Switch-Pak™, a general-purpose switch package, provides a complete solution for high level spill prevention with a fixed level switch point and 10 A compact relay controller for pump or valve actuation. These products are typically selected for corrosive bulk storage, day tank and waste sump applications.
These switches are available in 6" to 10-ft lengths, specified in 1/2" increments. Must specify length "A" when ordering.
$325.00 - $525.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Max Process Temperature (° C) Description
Availability Pricing
Flowline AV13-4213-A Single-point switch with strobe; sensor, float; wetted parts, PP; 120/240V EW-43305-10 Mfr # AV13-4213-A
COMPARE
150 80 Vertical-Mount Level Switch, Buoyancy Sensor, Polypropylene, Controller with Strobe Alert; 120/240 VAC, 50/60 Hz
$525.00USD / Each
Flowline AV16-4243-A Buoyancy Controller, junction box, SPDT reed, PP EW-68873-76 Mfr # AV16-4243-A
COMPARE
150 80 Vertical-Mount Level Switch, Buoyancy Sensor, Polypropylene, Junction Box with SPDT Reed; 12 to 36 VDC
$325.00USD / Each