Flowline EchoSpan Ultrasonic Level Transmitters (2-Wire)
  • Setup is fast and simple with push-button calibration and LCD display
  • Offered in three measurement ranges up to 10 m, with 2" dia transducer
  • 6-segment LCD display indicates level in inch or centimeter values
  • 7.6 cm minimum beam width for applications with restricted space
  • PC/ABS enclosure rated NEMA 4X with rugged PVDF transducer
  • Fail-safe intelligence with diagnostic feedback for easy troubleshooting
EchoSpan™, a general-purpose two-wire ultrasonic transmitter, provides noncontact level measurement up to 32 ft or 10 m, and is ideally suited for challenging ultrapure, corrosive or waste liquids. Push-button calibrated, the transmitter is broadly selected for atmospheric bulk storage, day tank and waste sump applications. Media examples include wastewater and sodium hydroxide.
$795.00 - $995.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
Flowline LU83-5101 Large Tank Transmitter, 4 to 20 mA, 8 EW-43260-00 Mfr # LU83-5101
COMPARE
26.2 30
$895.00USD / Each
Flowline LU81-5101 Large Tank Transmitter, 4 to 20 mA, 4 EW-43260-04 Mfr # LU81-5101
COMPARE
16.4 30
$795.00USD / Each
Flowline LU84-5101 Ultrasonic Level Transmitter; 32.8' Range, 2 EW-43312-65 Mfr # LU84-5101
COMPARE
32.8 30
$995.00USD / Each