Flowline Echotouch Intrinsically Safe Ultrasonic Level Transmitter (2-wire)
For general-purpose or hazardous applications
  • Intrinsically safe transmitter for use in hazardous environments
  • LCD digital display indicates level in inches or centimeters
  • EasyCal simple push-button calibration for all user set points
  • PP enclosure rated NEMA 4X with rugged PVDF transducer
  • Adjustable dead band and range filters eliminate false echo returns
CSA-approved and intrinsically safe for use in general-purpose or hazardous applications, the two-wire ultrasonic transmitter provides non-contact level measurement up to 18 ft or 5.4 m, and is ideally suited for challenging corrosive, slurry or waste liquids. Push-button calibrated, the transmitter is typically selected for atmospheric bulk storage, day tank and waste sump applications located within a classified area. Media examples include diesel fuel and sulfuric acid.
$850.00 - $950.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
Flowline LU20-5001-IS Ultrasonic Transmitter; 12 TO 32 VDC EW-43312-10 Mfr # LU20-5001-IS
COMPARE
18 30
$950.00USD / Each
Flowline LB11-1001 Intrinsically safe barrier; for use with intrinsically safe transmitters EW-43312-18 Mfr # LB11-1001
COMPARE
$850.00USD / Each