Flowline Liquid and Gas Flow Switches
Sturdy sensor withstands high flow velocities
  • Liquid switches are ideal for pump and process protection; gas switches for filter and process protection
  • Sensor, cable, and junction box are NEMA 4X rated—ideal for corrosive environments
  • FET switch output interfaces with PLC, SCADA, or alarms
  • Short sensors can be used in pipe diameters from 3/4" to 1 1/2"; long sensors from 2" to 16"
  • Easily adjust set point with a single turn of the adjustment knob
  • Switch state is selectable from normally open or normally closed
These general purpose liquid and gas flow switch provides reliable low or no-flow detection of clean, non-coating media with one 1A relay output. Available in Polypropylene-Ryton® and PVDF, the short flow sensor is used in pipe or ducting from 1/2” to 1 1/2”, and the long flow sensor is used in 2” and up. The flow switch set point may be adjusted from 0.04 to 3 fps in liquids, or 1 to 90 fps in gases as a low-flow alarm.
$200.00 - $345.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Cable Length (ft) Max Process Temperature (° C) Description
Availability Pricing
Flowline GT10-1305 Thermal Flow Switch for Gases, Short PP/Ryton Sensor EW-32756-00 Mfr # GT10-1305
COMPARE
10 60 Thermal Flow Switch for Gases, Short PP/Ryton Sensor, 3/4" NPT
$210.00USD / Each
Flowline GT10-5305 Thermal Flow Switch for Gases, Short PVDF Sensor EW-32756-02 Mfr # GT10-5305
COMPARE
10 60 Thermal Flow Switch for Gases, Short PVDF Sensor, 3/4" NPT
$335.00USD / Each
Flowline FT10-1425 Thermal Flow Switch for Liquids, Long PP/Ryton Sensor, 3/4" G EW-32756-03 Mfr # FT10-1425
COMPARE
10 60 Thermal Flow Switch for Liquids, Long PP/Ryton Sensor, 3/4" G
$215.00USD / Each
Flowline GT10-5405 Thermal Flow Switch for Gases, Long PVDF Sensor EW-32756-06 Mfr # GT10-5405
COMPARE
10 60 Thermal Flow Switch for Gases, Long PVDF Sensor, 3/4" NPT
$345.00USD / Each
Flowline FT10-1305 Thermal Flow Switch for Liquids, Short PP/Ryton Sensor EW-32756-08 Mfr # FT10-1305
COMPARE
10 60 Thermal Flow Switch for Liquids, Short PP/Ryton Sensor, 3/4" NPT
$200.00USD / Each
Flowline FT10-5305 Thermal Flow Switch for Liquids, Short PVDF Sensor EW-32756-10 Mfr # FT10-5305
COMPARE
10 60 Thermal Flow Switch for Liquids, Short PVDF Sensor, 3/4" NPT
$325.00USD / Each
Flowline FT10-1405 Thermal Flow Switch for Liquids, Long PP/Ryton Sensor EW-32756-12 Mfr # FT10-1405
COMPARE
10 60 Thermal Flow Switch for Liquids, Long PP/Ryton Sensor, 3/4" NPT
$210.00USD / Each
Flowline FT10-5405 Thermal Flow Switch for Liquids, Long PVDF Sensor EW-32756-14 Mfr # FT10-5405
COMPARE
10 60 Thermal Flow Switch for Liquids, Long PVDF Sensor, 3/4" NPT
$335.00USD / Each
Showing 1- 8 of 8