Flowline Nonintrusive RF Capacitance Level Switch
Measure level without touching fluid to avoid contamination
  • Ideal for applications where purity is important
If you are concerned about your liquid's purity, then this nonintrusive level switch is the solution. Sensor mounts on the outside of your nonmetallic tank and never touches your liquid. Use with clean, noncoating, nonscaling, and conductive liquids. NEMA 4X polysulfone sensor enclosure is ideal for industrial environments. Includes 8-ft cable. Weld mounting bracket (sold separately) in place for firm hold.
$230.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Process Temperature (° C) Description
Availability Pricing
Flowline LP50-1005 Non-intrusive RF Capacitance Level Switch, PP Bracket EW-43299-05 Mfr # LP50-1005
COMPARE
80 Non-intrusive RF Capacitance Level Switch, PP Bracket
$230.00USD / Each
Flowline LP50-6005 Non-intrusive RF Capacitance Level Switch, PE Bracket EW-43299-12 Mfr # LP50-6005
COMPARE
80 Non-intrusive RF Capacitance Level Switch, PE Bracket
$230.00USD / Each