Flowline Optic Leak Detection Switches
Detects leaks in and around secondary containment areas
  • Choose between polypropylene (PP) or PFA construction
The optic switch is an excellent choice for leak detection applications in and around the secondary containment vessel of sumps, tanks and pipes. The sensor can be applied in a broad range of liquids including sulfuric acid, jet fuel and de-ionized water. The relay output provides a reliable switch interface with remote devices such as a PLC, SCADA or alarm. For maximum flexibility, the sensor is offered in general purpose and intrinsically safe versions with two probe lengths and PP or PFA PTFE materials.
$175.00 - $295.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flowline LO10-1305 Optic Leak Detection Pp Sensor Short 12-36VDC EW-43312-01 Mfr # LO10-1305
COMPARE
Optic Leak Detection Pp Sensor Short 12-36VDC
$175.00USD / Each
Flowline LO10-1405 Optic Leak Detection Pp Sensor Long 12-36VDC EW-43312-02 Mfr # LO10-1405
COMPARE
Optic Leak Detection Pp Sensor Long 12-36VDC
$185.00USD / Each
Flowline LO10-2305 Optic Leak Detection Pfa Sensor Short 12-36VDC EW-43312-03 Mfr # LO10-2305
COMPARE
Optic Leak Detection Pfa Sensor Short 12-36VDC
$295.00USD / Each