Flowline Plastic Mini-Float Level Switches
Control liquid level even in corrosive environments
  • Excellent for clean, non-coating or non-scaling liquids
Choose either vertical or horizontal mounting to fit your application. Reliable reed switch output for interface with PLC or relay control devices. Your liquid must have a specific gravity of 0.8 or greater.
$28.00 - $56.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Max Process Temperature (° C) Description
Availability Pricing
Flowline LV20-1201 Mini-Float Level Switch, Polypropylene, 1/8" NPT(M) EW-43299-24 Mfr # LV20-1201
COMPARE
25 90 Float level switches; PP; 1/8" NPT(M)
$28.00USD / Each
Flowline LV20-5201 Mini-Float PVDF Level Switch, SPST, 1/8"NPT(M) EW-43299-26 Mfr # LV20-5201
COMPARE
25 90 Mini-Float PVDF Level Switch, SPST, 1/8"NPT(M)
$56.00USD / Each