Flowline Switch-Pro Remote Relay Level Controller
Control large loads—SPDT relay(s) rated 250 VAC, 10 A resistive
  • LED indicators provide user with power, sensor and relay status
  • Fail-safe relay control of pumps and valves with 0-60 second time delay
  • PP 35 mm DIN enclosure with removable terminal straps
Controllers accept all Flowline(R) point level sensors and allow you to control external devices like pumps, valves, or alarms. Adjustable time delay prevents frequent relay-cycling. LEDs indicate wet/dry sensor condition. Mount controllers on any 35-mm DIN rail.
$210.00 - $295.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Process Temperature (° C) Description
Availability Pricing
Flowline LC40-1001 Remote Relay Level Controller; one sensor, one relay channel EW-43300-30 Mfr # LC40-1001
COMPARE
70 Remote Relay Level Controller; one sensor, one relay channel
$210.00USD / Each
Flowline LC41-1001 Remote Relay Level Controller; two sensors, one relay EW-43300-32 Mfr # LC41-1001
COMPARE
70 Remote Relay Level Controller; two sensors, one relay
$245.00USD / Each
Flowline LC42-1001 Remote Level Controller; 2 SPDT, 3 Sensor EW-43300-34 Mfr # LC42-1001
COMPARE
70 Remote Level Controller; 2 SPDT, 3 Sensor
$295.00USD / Each