Flowline Switch-Tek Ultrasonic Level Switches
Compatible with Smart Trak and Switch-Pak installation fittings
  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.
$285.00 - $415.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
Flowline LU10-1305 Ultrasonic Level Switch-PP, Short, 3/4" NPT EW-23002-14 Mfr # LU10-1305
COMPARE
150
$285.00USD / Each
Flowline LU10-1405 Ultrasonic Level Switch-PP, Long, 3/4" NPT EW-23002-15 Mfr # LU10-1405
COMPARE
150
$295.00USD / Each
Flowline LU10-2305 Ultrasonic Level Switch-PFA, Short, 3/4" NPT EW-23002-16 Mfr # LU10-2305
COMPARE
150
$405.00USD / Each
Flowline LU10-1305 Ultrasonic level switch; material, PP; mounting threads, 3/4" NPT (M) EW-43299-02 Mfr # LU10-1305
COMPARE
150
$285.00USD / Each
Flowline LU10-2305 Ultrasonic level switch; SPDT relay: 1 A at 60 VDC/VAC; 12-36 VDC EW-43299-06 Mfr # LU10-2305
COMPARE
150
$405.00USD / Each
Flowline LU10-2405 Ultrasonic level switch; material, PFA; mounting threads, 3/4" NPT (M) EW-43299-08 Mfr # LU10-2405
COMPARE
150
$415.00USD / Each