This product is now discontinued, View Related Products
Fluke 1630 and 1621 Ground Resistance Testers
Fluke 1630
  • Quick and easy use - no earth ground stakes are necessary
  • Large, 35mm (1.35 inch) jaw opening
  • Measures ground resistance from 0.025 to 1500 ø
  • Measures ground leakage current from 0.2 to 30mA
  • High and Low alarming
  • Automatic self calibration
  • Rugged carrying case and resistance check loop included
FL1621
  • 3-pole Fall of Potential earth testing for basic measurements
  • 2 pole resistance measurements for added versatility
  • Easily capture values with single button operation
  • Available for sale in the United States and Canada only
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
Fluke 1621 Resistance Earth Ground Tester EW-20021-74 Mfr # FLUKE-1621
COMPARE
Call For Price
Fluke 1621 KIT Earth Ground Testing Kit EW-20033-79 Mfr # FLUKE-1621 KIT
COMPARE
Call For Price
Fluke 1630 Stakeless Earth Ground Clamp Meter EW-26048-88 Mfr # FLUKE-1630